accessibility menu, dialog, popup

UserName

CAS RN 4471-05-0

IUPAC Name: 1-phenylhexan-1-ol
Synonyms: 1-phenylhexan-1-ol
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: SVCRDVHXRDRHCP-UHFFFAOYNA-N
SMILES: CCCCCC(O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results