missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclohexanecarboxylic Acid 95.0+%, TCI America™

Catalog No. p-7135444
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21801G 1 g
P21805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21801G Supplier TCI America Supplier No. P21801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1135-67-7
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00037152
InChI Key QXXHHHWXFHPNOS-UHFFFAOYSA-N
Synonym 1-phenylcyclohexanecarboxylic acid, 1-phenyl-1-cyclohexanecarboxylic acid, 1-phenyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-phenyl, acmc-1c6ij, 1-phenylcyclohexanecarboxylicacid, qxxhhhwxfhpnos-uhfffaoysa, 1-phenylcyclohexanecarboxylic acid #, cyclohexanecarboxylicacid, 1-phenyl, 1-phenyl-1-cyclohexane carboxylic acid
PubChem CID 70817
IUPAC Name 1-phenylcyclohexane-1-carboxylic acid
SMILES C1CCC(CC1)(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 123°C
Quantity 1 g
Formula Weight 204.27
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-1-cyclohexanecarboxylic Acid