accessibility menu, dialog, popup

UserName

CAS RN 1135-67-7

IUPAC Name: 1-phenylcyclohexane-1-carboxylic acid
Synonyms: 1-phenylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: QXXHHHWXFHPNOS-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCCCC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)