missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1-cyclohexanecarboxylic acid, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 170750500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 1-Phenyl-1-cyclohexanecarboxylic acid | |
| 1135-67-7 | |
| 100.0 | |
| 95% | |
| C13H16O2 | |
| MFCD00037152 | |
| 1-phenylcyclohexanecarboxylic acid, 1-phenyl-1-cyclohexanecarboxylic acid, 1-phenyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-phenyl, acmc-1c6ij, 1-phenylcyclohexanecarboxylicacid, qxxhhhwxfhpnos-uhfffaoysa, 1-phenylcyclohexanecarboxylic acid #, cyclohexanecarboxylicacid, 1-phenyl, 1-phenyl-1-cyclohexane carboxylic acid | |
| C1CCC(CC1)(C2=CC=CC=C2)C(=O)O | |
| 204.27 | |
| 204.27 |
| 95% | |
| 94.0 | |
| Authentic | |
| Glass bottle | |
| C6H5C6H10CO2H | |
| 50g | |
| QXXHHHWXFHPNOS-UHFFFAOYSA-N | |
| 1-phenylcyclohexane-1-carboxylic acid | |
| 70817 | |
| 95% |
Chemical Identifiers
| 1135-67-7 | |
| 204.27 | |
| QXXHHHWXFHPNOS-UHFFFAOYSA-N | |
| 70817 | |
| C1CCC(CC1)(C2=CC=CC=C2)C(=O)O |
| C13H16O2 | |
| MFCD00037152 | |
| 1-phenylcyclohexanecarboxylic acid, 1-phenyl-1-cyclohexanecarboxylic acid, 1-phenyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-phenyl, acmc-1c6ij, 1-phenylcyclohexanecarboxylicacid, qxxhhhwxfhpnos-uhfffaoysa, 1-phenylcyclohexanecarboxylic acid #, cyclohexanecarboxylicacid, 1-phenyl, 1-phenyl-1-cyclohexane carboxylic acid | |
| 1-phenylcyclohexane-1-carboxylic acid |
Safety and Handling
EINECSNumber : 214-495-8
TSCA : TSCA
RUO â Research Use Only