missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclohexanecarboxylic acid, 95%

Catalog No. AC170750100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170750100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC170750100 Supplier Thermo Scientific Chemicals Supplier No. 170750100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1135-67-7 | C13H16O2 | 204.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1135-67-7
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.27
MDL Number MFCD00037152
InChI Key QXXHHHWXFHPNOS-UHFFFAOYSA-N
Synonym 1-phenylcyclohexanecarboxylic acid, 1-phenyl-1-cyclohexanecarboxylic acid, 1-phenyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-phenyl, acmc-1c6ij, 1-phenylcyclohexanecarboxylicacid, qxxhhhwxfhpnos-uhfffaoysa, 1-phenylcyclohexanecarboxylic acid #, cyclohexanecarboxylicacid, 1-phenyl, 1-phenyl-1-cyclohexane carboxylic acid
PubChem CID 70817
IUPAC Name 1-phenylcyclohexane-1-carboxylic acid
SMILES C1CCC(CC1)(C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 119°C to 124°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula C6H5C6H10CO2H
Quantity 10 g
Formula Weight 204.27
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1-Phenyl-1-cyclohexanecarboxylic acid, 95%
Show More Show Less
EINECSNumber 214-495-8
TSCA TSCA

RUO – Research Use Only