Learn More
1-Phenyl-1-butyne, 98%
CAS: 622-76-4 | C10H10 | 130.19 g/mol
Supplier: Thermo Scientific Chemicals A1133209
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Phenyl-1-butyne | |
| 0.918 | |
| 68°C (154°F) | |
| CH3CH2C≡CC6H5 | |
| MFCD00039945 | |
| 1903397 | |
| FFFMSANAQQVUJA-UHFFFAOYSA-N | |
| but-1-ynylbenzene | |
| 69328 | |
| 98% |
| 622-76-4 | |
| 201°C to 203°C | |
| C10H10 | |
| 1.552 | |
| 10 g | |
| 1-phenyl-1-butyne, 1-butynylbenzene, benzene, 1-butynyl, 1-butyne, 1-phenyl, but-1-yn-1-ylbenzene, 1-but-1-ynyl benzene, 1-ethyl-2-phenylacetylene, phenylbutyne, ethylphenylacetylene, phenylethylacetylene | |
| CCC#CC1=CC=CC=C1 | |
| 130.19 | |
| 130.19 |
Chemical Identifiers
| 622-76-4 | |
| 130.19 | |
| FFFMSANAQQVUJA-UHFFFAOYSA-N | |
| 69328 | |
| CCC#CC1=CC=CC=C1 |
| C10H10 | |
| MFCD00039945 | |
| 1-phenyl-1-butyne, 1-butynylbenzene, benzene, 1-butynyl, 1-butyne, 1-phenyl, but-1-yn-1-ylbenzene, 1-but-1-ynyl benzene, 1-ethyl-2-phenylacetylene, phenylbutyne, ethylphenylacetylene, phenylethylacetylene | |
| but-1-ynylbenzene |
Safety and Handling
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 210-752-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only