missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-butyne, 98%

Catalog No. p-7081382
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1133206 5 g
AAA1133209 10 g
AAA1133214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1133206 Supplier Thermo Scientific Chemicals Supplier No. A1133206
Only null left

CAS: 622-76-4 | C10H10 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-76-4
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00039945
InChI Key FFFMSANAQQVUJA-UHFFFAOYSA-N
Synonym 1-phenyl-1-butyne, 1-butynylbenzene, benzene, 1-butynyl, 1-butyne, 1-phenyl, but-1-yn-1-ylbenzene, 1-but-1-ynyl benzene, 1-ethyl-2-phenylacetylene, phenylbutyne, ethylphenylacetylene, phenylethylacetylene
PubChem CID 69328
IUPAC Name but-1-ynylbenzene
SMILES CCC#CC1=CC=CC=C1

Specifications

Density 0.918
Boiling Point 201°C to 203°C
Flash Point 68°C (154°F)
Refractive Index 1.552
Linear Formula CH3CH2C≡CC6H5
Quantity 5 g
Beilstein 1903397
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 1-Phenyl-1-butyne
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-752-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only