missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,4-pentanedione, 96%

Catalog No. AAL1315103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1315103 1 g
AAL1315106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1315103 Supplier Thermo Scientific Chemicals Supplier No. L1315103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 583-05-1 | C11H12O2 | 176.215 g/mol

1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 583-05-1
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00092889
InChI Key RBLXWIPBPPVLPU-UHFFFAOYSA-N
Synonym 1-phenyl-1,4-pentanedione, 1-phenyl-pentane-1,4-dione, 1,4-pentanedione, 1-phenyl, acmc-20aosh, 1-phenyl-1,4-pentandione, 5-phenyl-2,5-pentanedione, benzil-related compound, 41, 1-phenyl pentane-1,4-dione
PubChem CID 136371
IUPAC Name 1-phenylpentane-1,4-dione
SMILES CC(=O)CCC(=O)C1=CC=CC=C1

Specifications

Boiling Point 158°C to 162°C (12 mmHg)
Refractive Index 1.533
Quantity 1 g
Beilstein 908230
Solubility Information Not miscible in water.
Formula Weight 176.22
Percent Purity 96%
Chemical Name or Material 1-Phenyl-1,4-pentanedione
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only