CAS RN 583-05-1
IUPAC Name:
1-phenylpentane-1,4-dione
Synonyms:
1-phenylpentane-1,4-dione
Molecular Weight (g/mol):
176.22
Molecular Formula:
C11H12O2
InChi Key:
RBLXWIPBPPVLPU-UHFFFAOYSA-N
SMILES:
CC(=O)CCC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1-Phenyl-1,4-pentanedione, 96%
1-Phenyl-1,4-pentanedione, 96%
CAS: 583-05-1 Molecular Formula: C11H12O2 Molecular Weight (g/mol): 176.215 MDL Number: MFCD00092889 InChI Key: RBLXWIPBPPVLPU-UHFFFAOYSA-N Synonym: 1-phenyl-1,4-pentanedione,1-phenyl-pentane-1,4-dione,1,4-pentanedione, 1-phenyl,acmc-20aosh,1-phenyl-1,4-pentandione,5-phenyl-2,5-pentanedione,benzil-related compound, 41,1-phenyl pentane-1,4-dione PubChem CID: 136371 IUPAC Name: 1-phenylpentane-1,4-dione SMILES: CC(=O)CCC(=O)C1=CC=CC=C1