missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol, 97%

Catalog No. p-7035710
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5343506 5 g
AAH5343514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5343506 Supplier Thermo Scientific Chemicals Supplier No. H5343506
Only null left

CAS: 214746-69-7 | C20H21NO | 291.394 g/mol

1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 214746-69-7
Molecular Formula C20H21NO
Molecular Weight (g/mol) 291.394
MDL Number MFCD17677347
InChI Key AIBIEAJBGHJQJF-UHFFFAOYSA-N
Synonym 1-phenyl-1-4-1-piperidinyl phenyl-2-propyn-1-ol, 1-phenyl-1-4-piperidin-1-yl phenyl prop-2-yn-1-ol, 1-phenyl-1-4-piperidinophenyl prop-2-yn-1-ol, 1-4-piperidinophenyl-1-phenyl-2-propyne-1-ol
PubChem CID 15536835
IUPAC Name 1-phenyl-1-(4-piperidin-1-ylphenyl)prop-2-yn-1-ol
SMILES C#CC(C1=CC=CC=C1)(C2=CC=C(C=C2)N3CCCCC3)O

Specifications

Melting Point 142°C to 144°C
Quantity 5 g
Formula Weight 291.39
Percent Purity 97%
Chemical Name or Material 1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only