CAS RN 214746-69-7
IUPAC Name:
1-phenyl-1-[4-(piperidin-1-yl)phenyl]prop-2-yn-1-ol
Synonyms:
1-phenyl-1-[4-(piperidin-1-yl)phenyl]prop-2-yn-1-ol
Molecular Weight (g/mol):
291.39
Molecular Formula:
C20H21NO
InChi Key:
AIBIEAJBGHJQJF-UHFFFAOYNA-N
SMILES:
OC(C#C)(C1=CC=CC=C1)C1=CC=C(C=C1)N1CCCCC1
1
–
1
of
1
results
1
Promotions Available
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol, 97%
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol, 97%
CAS: 214746-69-7 Molecular Formula: C20H21NO Molecular Weight (g/mol): 291.394 MDL Number: MFCD17677347 InChI Key: AIBIEAJBGHJQJF-UHFFFAOYSA-N Synonym: 1-phenyl-1-4-1-piperidinyl phenyl-2-propyn-1-ol,1-phenyl-1-4-piperidin-1-yl phenyl prop-2-yn-1-ol,1-phenyl-1-4-piperidinophenyl prop-2-yn-1-ol,1-4-piperidinophenyl-1-phenyl-2-propyne-1-ol PubChem CID: 15536835 IUPAC Name: 1-phenyl-1-(4-piperidin-1-ylphenyl)prop-2-yn-1-ol SMILES: C#CC(C1=CC=CC=C1)(C2=CC=C(C=C2)N3CCCCC3)O