missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,2-propanedione 97.0+%, TCI America™

Catalog No. P021010G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P021010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P021010G Supplier TCI America Supplier No. P021010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 579-07-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00008755
InChI Key BVQVLAIMHVDZEL-UHFFFAOYSA-N
Synonym 1-phenyl-1,2-propanedione, acetylbenzoyl, acetyl benzoyl, benzoylacetyl, methylphenylglyoxal, pyruvophenone, 1,2-propanedione, 1-phenyl, phenylmethyldiketone, benzoyl methyl ketone, methyl phenyl diketone
PubChem CID 11363
ChEBI CHEBI:63552
IUPAC Name 1-phenylpropane-1,2-dione
SMILES CC(=O)C(=O)C1=CC=CC=C1

Specifications

Boiling Point 217°C
Quantity 10 g
UN Number 1224
Formula Weight 148.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1,2-propanedione