missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,2,3-butanetrione 2-oxime, 98+%

Catalog No. p-7046992
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1707303 1 g
AAL1707306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1707303 Supplier Thermo Scientific Chemicals Supplier No. L1707303
Only null left

CAS: 6797-44-0 | C10H9NO3 | 191.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6797-44-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00466576
InChI Key JVPIIQSFMRUIAP-PKNBQFBNSA-N
Synonym 1-phenyl-1,2,3-butanetrione 2-oxime, 1,2,3-butanetrione, 1-phenyl-, 2-oxime, 1-phenylbutane-1,2,3-trione 2-oxime, 2-hydroxyimino-1-phenylbutane-1,3-dione, 3-isonitrosobenzoylacetone, 2-hydroxyimino-1-phenyl-1,3-butanedione, 2e-1-phenyl-1,2,3-butanetrione 2-oxime #, 2z-2-n-hydroxyimino-1-phenylbutane-1,3-dione
PubChem CID 9576558
IUPAC Name (2E)-2-hydroxyimino-1-phenylbutane-1,3-dione
SMILES CC(=O)C(=NO)C(=O)C1=CC=CC=C1

Specifications

Melting Point 128°C to 130°C
Odor Odorless
Quantity 1 g
Beilstein 1946580
Formula Weight 191.19
Percent Purity ≥98%
Chemical Name or Material 1-Phenyl-1,2,3-butanetrione 2-oxime
Hazard Category H301
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 229-869-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only