missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98.0+%, TCI America™
Supplier: TCI America P20565G
Specifications
| 1-Phenyl-1,2,3,4-tetrahydroisoquinoline | |
| 100°C | |
| C15H15N | |
| 5 g | |
| PRTRSEDVLBBFJZ-UHFFFAOYSA-N | |
| 1-phenyl-1,2,3,4-tetrahydroisoquinoline | |
| 100137 | |
| ≥98.0% (GC,T) |
| 22990-19-8 | |
| White | |
| MFCD02179241 | |
| 1,2,3,4-Tetrahydro-1-phenylisoquinoline | |
| C1CNC(C2=CC=CC=C21)C3=CC=CC=C3 | |
| 209.292 | |
| 209.29 | |
| Crystalline Powder |
Chemical Identifiers
| 22990-19-8 | |
| 209.292 | |
| PRTRSEDVLBBFJZ-UHFFFAOYSA-N | |
| 100137 | |
| C1CNC(C2=CC=CC=C21)C3=CC=CC=C3 |
| C15H15N | |
| MFCD02179241 | |
| 1,2,3,4-Tetrahydro-1-phenylisoquinoline | |
| 1-phenyl-1,2,3,4-tetrahydroisoquinoline |
Safety and Handling
TSCA : No