CAS RN 22990-19-8
CAS RN 22990-19-8
IUPAC Name:
1-phenyl-1,2,3,4-tetrahydroisoquinoline
Synonyms:
1-phenyl-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol):
209.29
Molecular Formula:
C15H15N
InChi Key:
PRTRSEDVLBBFJZ-UHFFFAOYNA-N
SMILES:
C1CC2=CC=CC=C2C(N1)C1=CC=CC=C1
1
–
1
of
1
results
1
1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98.0+%, TCI America™
CAS: 22990-19-8 Molecular Formula: C15H15N Molecular Weight (g/mol): 209.292 MDL Number: MFCD02179241 InChI Key: PRTRSEDVLBBFJZ-UHFFFAOYSA-N Synonym: 1,2,3,4-Tetrahydro-1-phenylisoquinoline PubChem CID: 100137 IUPAC Name: 1-phenyl-1,2,3,4-tetrahydroisoquinoline SMILES: C1CNC(C2=CC=CC=C21)C3=CC=CC=C3