missing translation for 'onlineSavingsMsg'
Learn More

1-Phenoxy-2-propanol, tech. 85%

Catalog No. AAL0459618
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0459618 50 g
AAL0459630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0459618 Supplier Thermo Scientific Chemicals Supplier No. L0459618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 770-35-4 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 770-35-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016861
InChI Key IBLKWZIFZMJLFL-UHFFFAOYNA-N
Synonym 1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312
PubChem CID 92839
IUPAC Name 1-phenoxypropan-2-ol
SMILES CC(O)COC1=CC=CC=C1

Specifications

Density 1.06
Boiling Point 242°C to 244°C
Flash Point 120°C (248°F)
Refractive Index 1.523
Quantity 50 g
Beilstein 1941356
Formula Weight 152.19
Percent Purity 85%
Grade Technical
Chemical Name or Material 1-Phenoxy-2-propanol
EINECSNumber 212-222-7
RTECSNumber UB8886500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only