missing translation for 'onlineSavingsMsg'
Learn More

1-Phenoxy-2-propanol 95.0+%, TCI America™

Catalog No. P011825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P011825ML 25 mL
P0118500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P011825ML Supplier TCI America Supplier No. P011825ML

Chemical Identifiers

CAS 770-35-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016861
InChI Key IBLKWZIFZMJLFL-UHFFFAOYNA-N
Synonym 1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312
PubChem CID 92839
IUPAC Name 1-phenoxypropan-2-ol
SMILES CC(O)COC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 233°C
Quantity 25 mL
Formula Weight 152.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenoxy-2-propanol
RTECSNumber UB8886500
TSCA Yes