missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyn-3-ol 97.0+%, TCI America™

Catalog No. P006910G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P006910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P006910G Supplier TCI America Supplier No. P006910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4187-86-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00004572
InChI Key LBSKEFWQPNVWTP-UHFFFAOYSA-N
Synonym 1-pentyn-3-ol, ethylethynylcarbinol, ethyl ethynyl carbinol, 1-pentyne-3-ol, 1-pentine-3-ol, 3-hydroxy-1-pentine, ethynyl-ethyl carbinol, acmc-1agpc
PubChem CID 92981
IUPAC Name pent-1-yn-3-ol
SMILES CCC(C#C)O

Specifications

Color Yellow
Boiling Point 125°C
Quantity 10 g
UN Number 1986
Formula Weight 84.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Pentyn-3-ol
RTECSNumber SC4758500
TSCA No