accessibility menu, dialog, popup

UserName

CAS RN 4187-86-4

IUPAC Name: pent-1-yn-3-ol
Synonyms: pent-1-yn-3-ol
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: LBSKEFWQPNVWTP-UHFFFAOYNA-N
SMILES: CCC(O)C#C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)