missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pentene 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 30°C |
| Quantity | 25 mL |
| UN Number | 1108 |
| Formula Weight | 70.14 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Pentene |
Safety and Handling
| EINECSNumber | (2)-0019 |
| RTECSNumber | SB2179000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |