missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene 98.0+%, TCI America™

Catalog No. P031625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P031625ML 25 mL
P0316500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P031625ML Supplier TCI America Supplier No. P031625ML

Chemical Identifiers

CAS 109-67-1
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00003567
InChI Key YWAKXRMUMFPDSH-UHFFFAOYSA-N
Synonym 1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
PubChem CID 8004
IUPAC Name pent-1-ene
SMILES CCCC=C

Specifications

Color Colorless
Boiling Point 30°C
Quantity 25 mL
UN Number 1108
Formula Weight 70.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Pentene
EINECSNumber (2)-0019
RTECSNumber SB2179000
TSCA Yes
Recommended Storage Refrigerator