missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene, 97%

Catalog No. AAA1672314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1672314 25 g
AAA1672322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1672314 Supplier Thermo Scientific Chemicals Supplier No. A1672314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 109-67-1 | C5H10 | 70.14 g/mol

1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, reducing agents, halogens and acids.

Chemical Identifiers

CAS 109-67-1
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00003567
InChI Key YWAKXRMUMFPDSH-UHFFFAOYSA-N
Synonym 1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
PubChem CID 8004
IUPAC Name pent-1-ene
SMILES CCCC=C

Specifications

Melting Point -138°C
Density 0.64
Boiling Point 30°C to 31°C
Flash Point −51°C (−60°F)
Refractive Index 1.3715
Quantity 25 g
UN Number UN1108
Beilstein 1731629
Merck Index 14,7122
Solubility Information Miscible with ethanol,ethyl ether,benzene and organic solvents. Immiscible with water.
Formula Weight 70.14
Percent Purity 97%
Chemical Name or Material 1-Pentene
Show More Show Less
Hazard Category H224-H304
Hazard Statement GHS H Statement
H224-H304
Extremely flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: 1-PENTENE
EINECSNumber 203-694-5
RTECSNumber SB2179000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only