Learn More
1-Penten-3-ol, 97%
CAS: 616-25-1 | C5H10O | 86.13 g/mol
Supplier: Thermo Scientific Chemicals B2015514
Description
1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
| 1-Penten-3-ol | |
| 0.838 | |
| 25°C (77°F) | |
| 1.425 | |
| 25 g | |
| 1719834 | |
| Slightly soluble in water. | |
| CCC(O)C=C | |
| 86.13 | |
| 86.13 |
| 616-25-1 | |
| 114°C to 115°C | |
| C5H10O | |
| MFCD00004573 | |
| UN1987 | |
| 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol | |
| VHVMXWZXFBOANQ-UHFFFAOYNA-N | |
| pent-1-en-3-ol | |
| 12020 | |
| ≥98% |
Chemical Identifiers
| 616-25-1 | |
| 86.13 | |
| VHVMXWZXFBOANQ-UHFFFAOYNA-N | |
| 12020 | |
| CCC(O)C=C |
| C5H10O | |
| MFCD00004573 | |
| 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol | |
| pent-1-en-3-ol |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 210-472-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only