accessibility menu, dialog, popup

UserName

CAS RN 616-25-1

IUPAC Name: pent-1-en-3-ol
Synonyms: 1-penten-3-ol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: VHVMXWZXFBOANQ-UHFFFAOYNA-N
SMILES: CCC(O)C=C
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)