missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pentadecene 95.0+%, TCI America™
$119.30 - $300.11
Chemical Identifiers
| CAS | 13360-61-7 |
|---|---|
| Molecular Formula | C15H30 |
| Molecular Weight (g/mol) | 210.405 |
| MDL Number | MFCD00008987 |
| InChI Key | PJLHTVIBELQURV-UHFFFAOYSA-N |
| Synonym | 1-pentadecene, pentadecene, pentadecene,1, unii-kh577lia74, ccris 5721, acmc-1bt6q |
| PubChem CID | 25913 |
| ChEBI | CHEBI:77506 |
| IUPAC Name | pentadec-1-ene |
| SMILES | CCCCCCCCCCCCCC=C |
Chemical Identifiers
| 13360-61-7 | |
| 210.405 | |
| PJLHTVIBELQURV-UHFFFAOYSA-N | |
| 25913 | |
| pentadec-1-ene |
| C15H30 | |
| MFCD00008987 | |
| 1-pentadecene, pentadecene, pentadecene,1, unii-kh577lia74, ccris 5721, acmc-1bt6q | |
| CHEBI:77506 | |
| CCCCCCCCCCCCCC=C |
Specifications
| 13360-61-7 | |
| Colorless | |
| C15H30 | |
| 5 mL | |
| PJLHTVIBELQURV-UHFFFAOYSA-N | |
| pentadec-1-ene | |
| 25913 | |
| 210.41 | |
| Liquid |
| -4°C | |
| 268°C | |
| MFCD00008987 | |
| 1-pentadecene, pentadecene, pentadecene,1, unii-kh577lia74, ccris 5721, acmc-1bt6q | |
| CCCCCCCCCCCCCC=C | |
| 210.405 | |
| CHEBI:77506 | |
| ≥95.0% (GC) | |
| 1-Pentadecene |
Safety and Handling
EINECSNumber : (2)-0027
TSCA : Yes