accessibility menu, dialog, popup

UserName

CAS RN 13360-61-7

IUPAC Name: pentadec-1-ene
Synonyms: 1-pentadecene
Molecular Weight (g/mol): 210.41
Molecular Formula: C15H30
InChi Key: PJLHTVIBELQURV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC=C
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)