missing translation for 'onlineSavingsMsg'
Learn More

1-Pentadecene 99.0+%, TCI America™

Catalog No. S03455ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03455ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03455ML Supplier TCI America Supplier No. S03455ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13360-61-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00008987
InChI Key PJLHTVIBELQURV-UHFFFAOYSA-N
Synonym 1-pentadecene, pentadecene, pentadecene,1, unii-kh577lia74, ccris 5721, acmc-1bt6q
PubChem CID 25913
ChEBI CHEBI:77506
IUPAC Name pentadec-1-ene
SMILES CCCCCCCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 268°C
Quantity 5 mL
Formula Weight 210.41
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Pentadecene [Standard Material for GC]
EINECSNumber (2)-0027
TSCA Yes