missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pentadecanol 95.0+%, TCI America™
Supplier: TCI America P0036100G
Specifications
| 1-Pentadecanol | |
| 44°C | |
| 133°C | |
| MFCD00004759 | |
| 1-pentadecanol, pentadecyl alcohol, pentadecanol, n-pentadecanol, n-1-pentadecanol, neodol 5, unii-333qva4g2q, dsstox_cid_7270, dsstox_rid_78381, dsstox_gsid_27270 | |
| CCCCCCCCCCCCCCCO | |
| 228.42 | |
| CHEBI:77468 | |
| ≥95.0% (GC) |
| 629-76-5 | |
| White | |
| C15H32O | |
| 100 g | |
| REIUXOLGHVXAEO-UHFFFAOYSA-N | |
| pentadecan-1-ol | |
| 12397 | |
| 228.42 | |
| Crystalline Lumps |
Chemical Identifiers
| 629-76-5 | |
| 228.42 | |
| REIUXOLGHVXAEO-UHFFFAOYSA-N | |
| 12397 | |
| pentadecan-1-ol |
| C15H32O | |
| MFCD00004759 | |
| 1-pentadecanol, pentadecyl alcohol, pentadecanol, n-pentadecanol, n-1-pentadecanol, neodol 5, unii-333qva4g2q, dsstox_cid_7270, dsstox_rid_78381, dsstox_gsid_27270 | |
| CHEBI:77468 | |
| CCCCCCCCCCCCCCCO |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes