missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(p-Tolyl)ethylamine 96.0+%, TCI America™
Supplier: TCI America T25051G
Specifications
| 1-(p-Tolyl)ethylamine | |
| Yellow | |
| C9H13N | |
| 1 g | |
| 4-(1-Aminoethyl)toluene | |
| CC(N)C1=CC=C(C)C=C1 | |
| 135.21 | |
| 135.21 | |
| Liquid |
| 586-70-9 | |
| 204°C | |
| MFCD02177111 | |
| 2735 | |
| UZDDXUMOXKDXNE-UHFFFAOYNA-N | |
| 1-(4-methylphenyl)ethan-1-amine | |
| 577386 | |
| ≥96.0% (GC,T) |
Chemical Identifiers
| 586-70-9 | |
| 135.21 | |
| UZDDXUMOXKDXNE-UHFFFAOYNA-N | |
| 577386 | |
| CC(N)C1=CC=C(C)C=C1 |
| C9H13N | |
| MFCD02177111 | |
| 4-(1-Aminoethyl)toluene | |
| 1-(4-methylphenyl)ethan-1-amine |
Safety and Handling
TSCA : No