missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(p-Tolyl)ethylamine 96.0+%, TCI America™
$50.05 - $146.31
Chemical Identifiers
| CAS | 586-70-9 |
|---|---|
| Molecular Formula | C9H13N |
| Molecular Weight (g/mol) | 135.21 |
| MDL Number | MFCD02177111 |
| InChI Key | UZDDXUMOXKDXNE-UHFFFAOYNA-N |
| Synonym | 4-(1-Aminoethyl)toluene |
| PubChem CID | 577386 |
| IUPAC Name | 1-(4-methylphenyl)ethan-1-amine |
| SMILES | CC(N)C1=CC=C(C)C=C1 |
Chemical Identifiers
| 586-70-9 | |
| 135.21 | |
| UZDDXUMOXKDXNE-UHFFFAOYNA-N | |
| 577386 | |
| CC(N)C1=CC=C(C)C=C1 |
| C9H13N | |
| MFCD02177111 | |
| 4-(1-Aminoethyl)toluene | |
| 1-(4-methylphenyl)ethan-1-amine |
Specifications
| 586-70-9 | |
| 204°C | |
| MFCD02177111 | |
| 2735 | |
| UZDDXUMOXKDXNE-UHFFFAOYNA-N | |
| 1-(4-methylphenyl)ethan-1-amine | |
| 577386 | |
| ≥96.0% (GC,T) | |
| 1-(p-Tolyl)ethylamine |
| Yellow | |
| C9H13N | |
| 1 g | |
| 4-(1-Aminoethyl)toluene | |
| CC(N)C1=CC=C(C)C=C1 | |
| 135.21 | |
| 135.21 | |
| Liquid |
Safety and Handling
TSCA : No