Learn More
1-(p-Toluenesulfonyl)indole, 95%
CAS: 31271-90-6 | C15H13NO2S | 271.33 g/mol
Supplier: Thermo Scientific Chemicals B2232006
Description
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 1-(p-Toluenesulfonyl)indole | |
| 84°C to 86°C | |
| MFCD01165032 | |
| 1-tosyl-1h-indole, n-tosylindole, 1-p-toluenesulfonyl indole, 1-4-methylbenzenesulfonyl indole, n-p-toluenesulfonyl indole, 1-4-methylphenyl sulfonyl-1h-indole, 1-tosylindole, 1-p-tolylsulfonyl indole, 1-p-toluenesulphonyl indole, 4-methylphenyl sulfonyl indole | |
| JNRRPYFLDADLJW-UHFFFAOYSA-N | |
| 1-(4-methylbenzenesulfonyl)-1H-indole | |
| 10956616 | |
| 95% |
| 31271-90-6 | |
| C15H13NO2S | |
| 5 g | |
| Insoluble in water. | |
| CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12 | |
| 271.33 | |
| 271.33 |
Chemical Identifiers
| 31271-90-6 | |
| 271.33 | |
| JNRRPYFLDADLJW-UHFFFAOYSA-N | |
| 10956616 |
| C15H13NO2S | |
| MFCD01165032 | |
| 1-tosyl-1h-indole, n-tosylindole, 1-p-toluenesulfonyl indole, 1-4-methylbenzenesulfonyl indole, n-p-toluenesulfonyl indole, 1-4-methylphenyl sulfonyl-1h-indole, 1-tosylindole, 1-p-tolylsulfonyl indole, 1-p-toluenesulphonyl indole, 4-methylphenyl sulfonyl indole | |
| CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only