missing translation for 'onlineSavingsMsg'
Learn More

1-Octylamine, 99%

CAS: 111-86-4 | C8H19N | 129.247 g/mol

$155.27 - $155.27

Chemical Identifiers

CAS 111-86-4
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008247
InChI Key IOQPZZOEVPZRBK-UHFFFAOYSA-N
Synonym octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
PubChem CID 8143
ChEBI CHEBI:7728
IUPAC Name octan-1-amine
SMILES CCCCCCCCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2419336
View Documents
Thermo Scientific Chemicals
B2419336
500 g
Each for $155.27
Only null left
 
Description

Description

1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

Solubility
Miscible with ethanol, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.
Specifications

Chemical Identifiers

111-86-4
129.247
IOQPZZOEVPZRBK-UHFFFAOYSA-N
8143
octan-1-amine
C8H19N
MFCD00008247
octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
CHEBI:7728
CCCCCCCCN

Specifications

111-86-4
0.78
60°C (144°F)
1.429
500 g
1679227
octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octan-1-amine
8143
129.25
1-Octylamine
-1°C
178°C to 179°C
C8H19N
MFCD00008247
UN2735
Air Sensitive
Miscible with ethanol,benzene and ethyl acetate. Slightly miscible with water.
CCCCCCCCN
129.247
CHEBI:7728
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H301+H311-H314-H332-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 203-916-0

RTECSNumber : RG8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only