Learn More
(1-Octyl)triphenylphosphonium bromide, 97%
CAS: 42036-78-2 | C26H32BrP | 455.42 g/mol
Supplier: Thermo Scientific Chemicals L0241214
Description
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
Specifications
| (1-Octyl)triphenylphosphonium bromide | |
| 61°C to 63°C | |
| CH3(CH2)7P(C6H5)3Br | |
| 25 g | |
| Hygroscopic | |
| OBLXVLWZBMAMHE-UHFFFAOYSA-M | |
| octyl(triphenyl)phosphanium;bromide | |
| 3084855 | |
| 97% |
| 42036-78-2 | |
| C26H32BrP | |
| MFCD00051874 | |
| 3642460 | |
| octyltriphenylphosphonium bromide, 1-octyl triphenylphosphonium bromide, n-octyl triphenylphosphonium bromide, n-octyltriphenylphosphonium bromide, octyltriphenylphosphanium bromide, acmc-1ao06, octyl triphenyl phosphanium bromide, n-octyl-triphenylphosphonium bromide, phosphonium,octyltriphenyl-, bromide 1:1, 1-octyl triphenyl phosphonium bromide | |
| [Br-].CCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 455.42 | |
| 455.42 |
Chemical Identifiers
| 42036-78-2 | |
| 455.42 | |
| OBLXVLWZBMAMHE-UHFFFAOYSA-M | |
| 3084855 | |
| [Br-].CCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C26H32BrP | |
| MFCD00051874 | |
| octyltriphenylphosphonium bromide, 1-octyl triphenylphosphonium bromide, n-octyl triphenylphosphonium bromide, n-octyltriphenylphosphonium bromide, octyltriphenylphosphanium bromide, acmc-1ao06, octyl triphenyl phosphanium bromide, n-octyl-triphenylphosphonium bromide, phosphonium,octyltriphenyl-, bromide 1:1, 1-octyl triphenyl phosphonium bromide | |
| octyl(triphenyl)phosphanium;bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 255-635-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only