missing translation for 'onlineSavingsMsg'
Learn More

(1-Octyl)triphenylphosphonium bromide, 97%

Catalog No. AAL0241206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0241206 5 g
AAL0241214 25 g
AAL0241222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0241206 Supplier Thermo Scientific Chemicals Supplier No. L0241206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42036-78-2 | C26H32BrP | 455.42 g/mol

(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.

Chemical Identifiers

CAS 42036-78-2
Molecular Formula C26H32BrP
Molecular Weight (g/mol) 455.42
MDL Number MFCD00051874
InChI Key OBLXVLWZBMAMHE-UHFFFAOYSA-M
Synonym octyltriphenylphosphonium bromide, 1-octyl triphenylphosphonium bromide, n-octyl triphenylphosphonium bromide, n-octyltriphenylphosphonium bromide, octyltriphenylphosphanium bromide, acmc-1ao06, octyl triphenyl phosphanium bromide, n-octyl-triphenylphosphonium bromide, phosphonium,octyltriphenyl-, bromide 1:1, 1-octyl triphenyl phosphonium bromide
PubChem CID 3084855
IUPAC Name octyl(triphenyl)phosphanium;bromide
SMILES [Br-].CCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 61°C to 63°C
Linear Formula CH3(CH2)7P(C6H5)3Br
Quantity 5 g
Beilstein 3642460
Sensitivity Hygroscopic
Formula Weight 455.42
Percent Purity 97%
Chemical Name or Material (1-Octyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-635-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only