missing translation for 'onlineSavingsMsg'
Learn More

1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%

Catalog No. p-176703
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC356760050 5 g Glass bottle
AC356760250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC356760050 Supplier Thermo Scientific Chemicals Supplier No. 356760050

CAS: 304680-36-2 | C12H23F6N2P | 340.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid

Chemical Identifiers

CAS 304680-36-2
Molecular Formula C12H23F6N2P
Molecular Weight (g/mol) 340.29
MDL Number MFCD03427617
InChI Key GRCIJNHHTXBJAK-UHFFFAOYSA-N
Synonym 1-methyl-3-n-octylimidazolium hexafluorophosphate, 3-methyl-1-octylimidazolium hexafluorophosphate, 1-methyl-3-octylimidazolium hexafluorophosphate, 1-octyl-3-methylimidazolium hexafluorophosphate, 1-methyl-3-n-octylimidazoliumhexafluorophosphate, acmc-20aj2z, dsstox_cid_27928, dsstox_rid_82681, dsstox_gsid_47952, ksc222c5l
PubChem CID 2734243
IUPAC Name 1-methyl-3-octylimidazol-1-ium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Yellow
Density 1.2345g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4230 to 1.4250
Quantity 5 g
Specific Gravity 1.2345
Viscosity 866 mPa.s (20°C)
Formula Weight 340.29
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Octyl-3-methylimidazolium hexafluorophosphate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only