accessibility menu, dialog, popup

UserName

CAS RN 304680-36-2

IUPAC Name: 3-methyl-1-octyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-octyl-3-methylimidazolium hexafluorophosphate
Molecular Weight (g/mol): 340.29
Molecular Formula: C12H23F6N2P
InChi Key: GRCIJNHHTXBJAK-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)