missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octen-3-ol 98.0+%, TCI America™
$82.79 - $248.34
Chemical Identifiers
| CAS | 3391-86-4 |
|---|---|
| Molecular Formula | C8H16O |
| Molecular Weight (g/mol) | 128.215 |
| MDL Number | MFCD00004589 |
| InChI Key | VSMOENVRRABVKN-UHFFFAOYSA-N |
| Synonym | 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol |
| PubChem CID | 18827 |
| ChEBI | CHEBI:34118 |
| IUPAC Name | oct-1-en-3-ol |
| SMILES | CCCCCC(C=C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O015925ML
|
TCI America
O015925ML |
25 mL |
Each for $82.79
|
|
|||||
|
O0159100ML
|
TCI America
O0159100ML |
100 mL |
Each for $248.34
|
|
|||||
Chemical Identifiers
| 3391-86-4 | |
| 128.215 | |
| VSMOENVRRABVKN-UHFFFAOYSA-N | |
| 18827 | |
| oct-1-en-3-ol |
| C8H16O | |
| MFCD00004589 | |
| 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol | |
| CHEBI:34118 | |
| CCCCCC(C=C)O |
Specifications
| 3391-86-4 | |
| 166°C | |
| MFCD00004589 | |
| 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol | |
| CCCCCC(C=C)O | |
| 128.215 | |
| CHEBI:34118 | |
| ≥98.0% (GC) | |
| 1-Octen-3-ol |
| Colorless | |
| C8H16O | |
| 25 mL | |
| VSMOENVRRABVKN-UHFFFAOYSA-N | |
| oct-1-en-3-ol | |
| 18827 | |
| 128.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2393
RTECSNumber : RH3300000
TSCA : Yes