missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octen-3-ol 98.0+%, TCI America™
Supplier: TCI America O0159100ML
Specifications
| 1-Octen-3-ol | |
| Colorless | |
| C8H16O | |
| 100 mL | |
| VSMOENVRRABVKN-UHFFFAOYSA-N | |
| oct-1-en-3-ol | |
| 18827 | |
| 128.22 | |
| Liquid |
| 3391-86-4 | |
| 166°C | |
| MFCD00004589 | |
| 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol | |
| CCCCCC(C=C)O | |
| 128.215 | |
| CHEBI:34118 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3391-86-4 | |
| 128.215 | |
| VSMOENVRRABVKN-UHFFFAOYSA-N | |
| 18827 | |
| oct-1-en-3-ol |
| C8H16O | |
| MFCD00004589 | |
| 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol | |
| CHEBI:34118 | |
| CCCCCC(C=C)O |
Safety and Handling
EINECSNumber : (2)-2393
RTECSNumber : RH3300000
TSCA : Yes