missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanol, Reagents
1-Octanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2257850400C2
| Quantity | 400 g |
|---|
Chemical Identifiers
| 111-87-5 | |
| 130.23 | |
| n-Octanol, n-Octyl Alcohol, normal Octanol | |
| CCCCCCCCO |
| C8H18O | |
| KBPLFHHGFOOTCA-UHFFFAOYSA-N | |
| octan-1-ol |
Specifications
| 1-Octanol | |
| 196°C | |
| Colorless | |
| -18°C | |
| C8H18O | |
| 0.827 | |
| KBPLFHHGFOOTCA-UHFFFAOYSA-N | |
| octan-1-ol | |
| Reagent |
| 111-87-5 | |
| -18°C | |
| 196°C | |
| Glass Bottle | |
| 400 g | |
| n-Octanol, n-Octyl Alcohol, normal Octanol | |
| CCCCCCCCO | |
| 130.23 | |
| Liquid |