missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanethiol 98.0+%, TCI America™
$42.87 - $106.25
Chemical Identifiers
| CAS | 111-88-6 |
|---|---|
| Molecular Formula | C8H18S |
| Molecular Weight (g/mol) | 146.29 |
| MDL Number | MFCD00004912 |
| InChI Key | KZCOBXFFBQJQHH-UHFFFAOYSA-N |
| Synonym | 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol |
| PubChem CID | 8144 |
| IUPAC Name | octane-1-thiol |
| SMILES | CCCCCCCCS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O002525ML
|
TCI America
O002525ML |
25 mL |
Each for $42.87
|
|
|||||
|
O0025500ML
|
TCI America
O0025500ML |
500 mL |
Each for $106.25
|
|
|||||
Chemical Identifiers
| 111-88-6 | |
| 146.29 | |
| KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
| 8144 | |
| CCCCCCCCS |
| C8H18S | |
| MFCD00004912 | |
| 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
| octane-1-thiol |
Specifications
| 111-88-6 | |
| 200°C | |
| MFCD00004912 | |
| 3082 | |
| KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
| octane-1-thiol | |
| 8144 | |
| ≥98.0% (GC) | |
| 1-Octanethiol |
| Yellow | |
| C8H18S | |
| 25 mL | |
| 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
| CCCCCCCCS | |
| 146.29 | |
| 146.29 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0464
TSCA : Yes