missing translation for 'onlineSavingsMsg'
Learn More

1-Octanethiol, 98%

CAS: 111-88-6 | C8H18S | 146.29 g/mol

Supplier:  Thermo Scientific Chemicals L07195AP

 View more versions of this product

Catalog No. AAL07195AP


Only null left
Explore available promotions

Description

Description

1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1-Octanethiol
-49°C
197°C to 199°C
C8H18S
MFCD00004912
UN3082
1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol
KZCOBXFFBQJQHH-UHFFFAOYSA-N
octane-1-thiol
8144
98%
111-88-6
0.843
68°C (154°F)
1.455
500 mL
1733101
Not miscible in water.
CCCCCCCCS
146.29
146.29

Chemical Identifiers

111-88-6
146.29
KZCOBXFFBQJQHH-UHFFFAOYSA-N
8144
CCCCCCCCS
C8H18S
MFCD00004912
1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol
octane-1-thiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H332-H317-H227
Harmful if inhaled.
May cause an allergic skin reaction.
Combustible liquid.

P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c

H227-H315-H317-H319

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 203-918-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only