Learn More
1-Octanethiol, 98%
CAS: 111-88-6 | C8H18S | 146.29 g/mol
Supplier: Thermo Scientific Chemicals L07195AE
Description
1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 1-Octanethiol | |
| -49°C | |
| 197°C to 199°C | |
| C8H18S | |
| MFCD00004912 | |
| UN3082 | |
| 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
| KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
| octane-1-thiol | |
| 8144 | |
| 98% |
| 111-88-6 | |
| 0.843 | |
| 68°C (154°F) | |
| 1.455 | |
| 100 mL | |
| 1733101 | |
| Not miscible in water. | |
| CCCCCCCCS | |
| 146.29 | |
| 146.29 |
Chemical Identifiers
| 111-88-6 | |
| 146.29 | |
| KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
| 8144 | |
| CCCCCCCCS |
| C8H18S | |
| MFCD00004912 | |
| 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
| octane-1-thiol |
Safety and Handling
GHS H Statement
H332-H317-H227
Harmful if inhaled.
May cause an allergic skin reaction.
Combustible liquid.
P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c
H227-H315-H317-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 203-918-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only