missing translation for 'onlineSavingsMsg'
Learn More

1-Octanethiol, 97%

CAS: 111-88-6 | C8H18S | 146.29 g/mol

Supplier:  Thermo Scientific Chemicals 157480020

Catalog No. AC157480020


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Octanethiol
-49.0°C
197.0°C to 200.0°C
Authentic
C8H18S
CH3(CH2)7SH
2 kg
1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol
CCCCCCCCS
146.29
146.29
111-88-6
0.8400g/mL
68°C
96% min. (GC)
1.4525 to 1.4545
MFCD00004912
0.84
KZCOBXFFBQJQHH-UHFFFAOYSA-N
octane-1-thiol
8144
97%

Chemical Identifiers

111-88-6
146.29
KZCOBXFFBQJQHH-UHFFFAOYSA-N
8144
CCCCCCCCS
C8H18S
MFCD00004912
1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol
octane-1-thiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 203-918-1

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only