Learn More
1-Octanethiol, 97%
CAS: 111-88-6 | C8H18S | 146.29 g/mol
Supplier: Thermo Scientific Chemicals 157480025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Octanethiol | |
| 96.0 | |
| Colorless | |
| Authentic | |
| Plastic Bottle | |
| 1.4525 to 1.4545 (20°C, 589nm) | |
| MFCD00004912 | |
| 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
| CCCCCCCCS | |
| 146.29 | |
| 146.29 | |
| Liquid |
| 111-88-6 | |
| 100.0 | |
| 0.8400g/mL | |
| 97% | |
| C8H18S | |
| CH3(CH2)7SH | |
| 2.5 kg | |
| KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
| octane-1-thiol | |
| 8144 | |
| ≥96.0% |
Chemical Identifiers
| 111-88-6 | |
| 146.29 | |
| KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
| 8144 | |
| CCCCCCCCS |
| C8H18S | |
| MFCD00004912 | |
| 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
| octane-1-thiol |
Safety and Handling
GHS P Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only