Learn More
1-Octanethiol, 97%
CAS: 111-88-6 | C8H18S | 146.29 g/mol
Supplier: Thermo Scientific Chemicals 157480010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Octanethiol | |
| 96.0 | |
| -49.0°C | |
| 0.8400g/mL | |
| 68°C | |
| 96% min. (GC) | |
| C8H18S | |
| CH3(CH2)7SH | |
| 1 kg | |
| 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
| CCCCCCCCS | |
| 146.29 | |
| 146.29 | |
| Liquid |
| 111-88-6 | |
| 100.0 | |
| Colorless | |
| 197.0°C to 200.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4525 to 1.4545 | |
| MFCD00004912 | |
| 0.84 | |
| KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
| octane-1-thiol | |
| 8144 | |
| 97% |
Chemical Identifiers
| 111-88-6 | |
| 146.29 | |
| KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
| 8144 | |
| CCCCCCCCS |
| C8H18S | |
| MFCD00004912 | |
| 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
| octane-1-thiol |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 203-918-1
RUO – Research Use Only