missing translation for 'onlineSavingsMsg'
Learn More

1-(o-Tolyl)piperazine 98.0+%, TCI America™

Catalog No. p-7137737
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34045G 5 g
T340425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34045G Supplier TCI America Supplier No. T34045G

Chemical Identifiers

CAS 39512-51-1
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD00040729
InChI Key WICKLEOONJPMEQ-UHFFFAOYSA-N
Synonym 1-2-methylphenyl piperazine, 1-o-tolyl piperazine, 4-2-methylphenyl piperazine, 1-o-tolylpiperazine, piperazine, 1-2-methylphenyl, 1-o-tolyl-piperazine, 1-2-methylphenyl-piperazine, o-tolylpiperazine, 2-methylphenylpiperazine, chembl9746
PubChem CID 91965
IUPAC Name 1-(2-methylphenyl)piperazine
SMILES CC1=CC=CC=C1N2CCNCC2

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 161°C
Quantity 5 g
UN Number 3263
Formula Weight 176.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(o-Tolyl)piperazine