missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-O-Galloyl-D-glucose, ≥90% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 13405-60-2 |
| Physical Form | Neat |
| Linear Formula | C13H16O10 |
| MDL Number | MFCD11975493 |
| Synonym | beta-D-Glucopyranose 1-(3,4,5-trihydroxybenzoate); beta-Glucogallin; 1-O-Galloyl-beta-D-glucopyranose |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 332.26 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Analytical Standard |