missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nonanol 99.0+%, TCI America™
$32.28 - $113.92
Chemical Identifiers
| CAS | 143-08-8 |
|---|---|
| Molecular Formula | C9H20O |
| Molecular Weight (g/mol) | 144.258 |
| MDL Number | MFCD00002990 |
| InChI Key | ZWRUINPWMLAQRD-UHFFFAOYSA-N |
| Synonym | 1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol |
| PubChem CID | 8914 |
| ChEBI | CHEBI:35986 |
| IUPAC Name | nonan-1-ol |
| SMILES | CCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
N029225ML
|
N/A | 25 mL |
Each for $32.28
|
|
|||||
|
N0292500ML
|
N/A | 500 mL |
Each for $113.92
|
|
|||||
Chemical Identifiers
| C9H20O | |
| MFCD00002990 | |
| 1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol | |
| CHEBI:35986 | |
| CCCCCCCCCO |
Specifications
| 143-08-8 | |
| 214°C | |
| MFCD00002990 | |
| 1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol | |
| CCCCCCCCCO | |
| 144.258 | |
| CHEBI:35986 | |
| ≥99.0% (GC) | |
| 1-Nonanol |
| Colorless | |
| C9H20O | |
| 3082 | |
| ZWRUINPWMLAQRD-UHFFFAOYSA-N | |
| nonan-1-ol | |
| 8914 | |
| 144.26 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : RB1575000
TSCA : Yes