missing translation for 'onlineSavingsMsg'
Learn More

1-Nonanol 99.0+%, TCI America™

Catalog No. N029225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N029225ML 25 mL
N0292500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N029225ML Supplier TCI America Supplier No. N029225ML

Chemical Identifiers

CAS 143-08-8
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00002990
InChI Key ZWRUINPWMLAQRD-UHFFFAOYSA-N
Synonym 1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
PubChem CID 8914
ChEBI CHEBI:35986
IUPAC Name nonan-1-ol
SMILES CCCCCCCCCO

Specifications

Color Colorless
Boiling Point 214°C
Quantity 25 mL
UN Number 3082
Formula Weight 144.26
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nonanol
EINECSNumber (2)-0217
RTECSNumber RB1575000
TSCA Yes