missing translation for 'onlineSavingsMsg'
Learn More

1-Nonanol 99.0+%, TCI America™

Supplier:  TCI America N0292500ML

 View more versions of this product

Catalog No. N0292500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Nonanol
Colorless
C9H20O
500 mL
1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
CCCCCCCCCO
144.258
CHEBI:35986
≥99.0% (GC)
143-08-8
214°C
MFCD00002990
3082
ZWRUINPWMLAQRD-UHFFFAOYSA-N
nonan-1-ol
8914
144.26
Liquid

Chemical Identifiers

143-08-8
144.258
ZWRUINPWMLAQRD-UHFFFAOYSA-N
8914
nonan-1-ol
C9H20O
MFCD00002990
1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
CHEBI:35986
CCCCCCCCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : RB1575000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions